Skip to Content
Merck
All Photos(1)

Documents

81210

Sigma-Aldrich

Poly(ethylene glycol)

average Mw 1,500

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol), average Mw 1,500

form

solid

Quality Level

mol wt

average Mw 1,500

mp

45-50 °C

transition temp

Tm 45-50 °C

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, required for the purification of biological materials. PEG also interacts with cell membrane, thereby resulting in cell fusion.

Application

Poly(ethylene glycol) has been used for histological, histochemical and immunohistochemical evaluation of in vitro cultured beads. It has been used for the fusion of cells.

Other Notes

Molecular weight range: 1,400-1,600

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chondrogenesis of human mesenchymal stem cells encapsulated in alginate beads.
Ma HL et al.
Journal of Biomedical Materials Research Part A, 64, 273-273 (2003)
DC-SIGN (CD209) expression is IL-4 dependent and is negatively regulated by IFN, TGF-beta, and anti-inflammatory agents.
Relloso M et al.
Journal of Immunology, 168, 2634-2634 (2002)
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Angewandte Chemie (International Edition in English), 49, 6288-6308 (2010)
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service