Skip to Content
Merck
All Photos(1)

Documents

49143

Supelco

Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate

traceable to primary standards (LGC)

Synonym(s):

Potassium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the label

mp

334 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mettler-Toledo calibration substance ME 51143095, potassium nitrate is provided with the melting point specific to Mettler-Toledo instrument, documented under both thermodynamic mode and pharmacopeia modes.
The melting point value equals a mean of 6 to 12 measurements with a Mettler-Toledo MP90 Excellence instrument that is calibrated with primary standards. The mp value is found using Capillary method, in accordance with European Pharmacopeia (2.2.14.)

Application

Potassium nitrate, Mettler-Toledo calibration substance ME 51143095 is used as a melting point calibration standard to assess the performance of Mettler-Toledo melting point instruments.

Features and Benefits

  • Melting point traceable to a primary standard (LGC, London)
  • Physically correct and heating rate dependent melting point evaluations using thermodynamic and pharmacopeia modes of testing, respectively
  • Certificates of analysis and safety data sheet provided
  • Uncertainty of measurement up to ± 0.5 °C

Legal Information

Mettler-Toledo is a registered trademark of Mettler-Toledo, Inc.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dengjun Wang et al.
Journal of contaminant hydrology, 164, 35-48 (2014-06-14)
The increasing application of engineered nanoparticles (ENPs) has heightened the concern that these ENPs would eventually be released to the environment and may enter into life cycle of living beings. In this regard, it is essential to understand how these
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Mila Ivanova Luleva et al.
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
M A Olutoye et al.
Bioresource technology, 102(23), 10777-10783 (2011-10-11)
Fatty acid methyl esters (FAME) were produced from palm oil using eggshell modified with magnesium and potassium nitrates to form a composite, low-cost heterogeneous catalyst for transesterification. The catalyst, prepared by the combination of impregnation/co-precipitation was calcined at 830 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service