Skip to Content
Merck
All Photos(1)

Key Documents

41131

Supelco

Mettler-Toledo Calibration substance ME 51143091, Saccharin

traceable to primary standards (LGC)

Synonym(s):

Saccharin, 2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

quality

traceable to primary standards (LGC)

shelf life

limited shelf life, expiry date on the label

mp

226-229 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mettler-Toledo calibration substance ME 51143091, saccharin is an analytical standard, designed for the routine calibration of Mettler-Toledo melting point instrument. Its value is an average of 6 to 12 measurements with a Mettler-Toledo MP90 Excellence instrument that is calibrated against primary standards. The melting point is validated by Capillary method as per European Pharmacopeia (2.2.14.)

Application

Saccharin Mettler-Toledo calibration substance ME 51143091 is a melting point calibration standard used to calibrate Mettler-Toledo mp instruments.

Biochem/physiol Actions

A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Features and Benefits

  • Traceable to a primary standard from LGC, London
  • Melting point evaluation conducted in both thermodynamic and pharmacopeia modes for physically correct and heating rate dependent melting point determinations, respectively
  • Available with certificates of analysis and safety data sheet
  • A product of analytical standard grade to help meet the QC/QA requirements of melting point determination
  • Uncertainty of measurement up to ± 0.3 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amjad Alhalaweh et al.
Molecular pharmaceutics, 9(9), 2605-2612 (2012-08-08)
Cocrystals constitute an important class of pharmaceutical solids for their remarkable ability to modulate solubility and pH dependence of water insoluble drugs. Here we show how cocrystals of indomethacin-saccharin (IND-SAC) and carbamazepine-saccharin (CBZ-SAC) enhance solubility and impart a pH-sensitivity different
Robert P Farrell et al.
Organic letters, 15(1), 168-171 (2012-12-14)
A process for the regioselective amination of unsymmetrical 3,5-substituted pyridine N-oxides has been developed utilizing cheap, readily available saccharin as an ammonium surrogate. High conversions of the corresponding saccharin adducts have been achieved under mild reaction conditions. In situ deprotection
Steven Zukerman et al.
Physiology & behavior, 109, 33-41 (2012-12-04)
In a recent study, intragastric (IG) self-infusion of 16% glucose stimulated 1-h intake and conditioned a preference for a flavored saccharin solution in C57BL/6J mice (Zukerman et al., 2011). Experiment 1 of the present study presents a concentration-response analysis of
Hellmuth Lilienthal et al.
Toxicology, 311(1-2), 52-60 (2013-02-14)
Many chemicals are known to exhibit endocrine activity and affect reproductive functions in vertebrates and invertebrates. Endocrine effects include influences on sexual differentiation of the brain during development and reproductive and non-reproductive behavior in adult offspring. We previously demonstrated that
Jennifer E Nyland et al.
Behavioural brain research, 240, 87-90 (2012-11-24)
Addiction is a chronic disease where periods of abstinence are riddled with instances of craving, withdrawal, and eventual relapse to escalated drug use. Cues previously associated with drug use can have a deleterious effect on this cycle by precipitating withdrawal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service