Skip to Content
Merck
All Photos(1)

Documents

46214

Supelco

3-Octanone

analytical standard

Synonym(s):

Ethyl amyl ketone, Ethyl pentyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COC2H5
CAS Number:
Molecular Weight:
128.21
Beilstein:
1700021
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.415 (lit.)

bp

167-168 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCCC(=O)CC

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h3-7H2,1-2H3

InChI key

RHLVCLIPMVJYKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Octanone may be used as an analytical reference standard for the quantification of the analyte in truffle aromas using gas chromatography (GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile organic compounds from truffles via SPME-GC-MS
Mauriello G, et al.
Journal of Chromatographic Science, 42(6), 299-305 (2004)
R N Sinha et al.
Mycopathologia, 101(1), 53-60 (1988-01-01)
The fungal odor compounds 3-methyl-1-butanol, 1-octen-3-ol and 3-octanone were monitored in nine experimental bins in Winnipeg, Manitoba containing a hard red spring wheat during the autumn, winter and summer seasons of 1984-85. Quality changes were associated with seed-borne microflora and
William O H Hughes et al.
Journal of economic entomology, 95(3), 537-543 (2002-06-22)
Leaf-cutting ants are important economic pests of the Neotropics, and the most common method of control involves the use of insecticidal baits. Baits that are currently available exhibit low attractiveness to grass-cutting species, thus there is a need to develop
Glen C Rains et al.
Biotechnology progress, 22(1), 2-8 (2006-02-04)
A portable, handheld volatile odor detector ("Wasp Hound") that utilizes a computer vision system and Microplitis croceipes (Cresson) (Hymenoptera: Braconidae), a parasitoid wasp, as the chemical sensor was created. Five wasps were placed in a test cartridge and placed inside
Hari N Pati et al.
European journal of medicinal chemistry, 44(1), 54-62 (2008-05-13)
This study demonstrated that replacement of the axial protons on the C2 and C6 atoms of various 1-methyl-3,5-bis(benzylidene)-4-piperidones 3 by a dimethylene bridge leading to series 2 lowered cytotoxic potencies. Four compounds 2a and 3a-c emerged as lead molecules based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service