All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H20N2O2
CAS Number:
Molecular Weight:
188.27
Beilstein:
2080745
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
CCCOC(=O)NCCCN(C)C
InChI
1S/C9H20N2O2/c1-4-8-13-9(12)10-6-5-7-11(2)3/h4-8H2,1-3H3,(H,10,12)
InChI key
WZZLDXDUQPOXNW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Propamocarb is a systemic fungicide, widely used for greenhouse vegetables and agricultural commodities, which shows protective action against phycomycetous diseases (Phythium, Phytophthora spp.).[1]
Application
Propamocarb may be used as an analytical reference standard for the quantification of the analyte in vegetables[1] and fruits samples[1][2] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
212.0 °F
Flash Point(C)
> 100 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eva C Bonefeld-Jorgensen et al.
Molecular and cellular endocrinology, 244(1-2), 20-30 (2005-10-13)
The estrogenic potential of four pesticides (endosulfan, prochloraz, tolchlofos-methyl and propamocarb) was compared in parallel with 17beta-estradiol (E2) by reporter constructs in transient transfected MCF-7BUS and in stable transfected MVLN cells. Similar detection limit and half maximum effect concentration was
Lin Ye et al.
Scientific reports, 10(1), 177-177 (2020-01-15)
The extensive use of chemical fertilizers poses serious collateral problems such as environmental pollution, pest resistance development and food safety decline. Researches focused on applying plant-beneficial microorganisms to partially replace chemical fertilizer use is increasing due to the requirement of
Rui Zhang et al.
Acta biochimica et biophysica Sinica, 51(1), 88-96 (2018-12-14)
Propamocarb (PM) is a pesticide that is widely used to protect cucumbers and other plants from downy mildew. Recently, some studies indicated that PM exposure had potential toxic effects in animals. In this study, adult male zebrafish were exposed to
H S Elsabbagh et al.
Pharmacological research, 43(1), 71-76 (2001-02-24)
Female mice received either cupravit or previcur at dose levels of 300 ppm and 1000 ppm, respectively, for 8 weeks. Humoral immune response was evaluated by measuring the plaque-forming cell (PFC) response and hemolysing antibody titers against sheep red blood
Sherif H Abd-Alrahman et al.
Bulletin of environmental contamination and toxicology, 89(2), 302-305 (2012-06-16)
Propamocarb-hydrochloride is a widely used fungicide around the world. The dissipation of propamocarb-hydrochloride in tomatoes, potatoes and cucumber were evaluated. QuEChERS methodology before quantification using HPLC-DAD was used. Preharvest interval and maximum residue limit were 4 days and 1.0 mg
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service