Skip to Content
Merck
All Photos(1)

Documents

34326

Supelco

Cymoxanil

PESTANAL®, analytical standard

Synonym(s):

1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N4O3
CAS Number:
Molecular Weight:
198.18
Beilstein:
2214018
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNC(=O)NC(=O)C(=N\OC)\C#N

InChI

1S/C7H10N4O3/c1-3-9-7(13)10-6(12)5(4-8)11-14-2/h3H2,1-2H3,(H2,9,10,12,13)/b11-5+

InChI key

XERJKGMBORTKEO-VZUCSPMQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cymoxanil is an agricultural fungicide effective against Peronosporales fungus affecting fruits and vegetables.

Application

Cymoxanil may be used as an analytical reference standard for the determination of the analyte in foodstuffs, soil, commercial pesticide formulation and tobacco by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Sensitive, handle under inert gas.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Blood,thymus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sensitivity to cymoxanil in populations of Plasmopara viticola in northern Italy.
Gullino ML, et al.
Plant Pathology, 46(5), 729-736 (1997)
Determination of simazine and cymoxanil in soils by microwave-assisted solvent extraction and HPLC with reductive amperometrical detection.
de Sabando OL, et al.
Chromatographia, 55(11-12), 667-671 (2002)
Rapid determination of famoxadone and cymoxanil in commercial pesticide formulation by high performance liquid chromatography using a C18 monolithic rod column.
Balayiannis G, et al.
Bulletin of Environmental Contamination and Toxicology, 93(6), 775-780 (2014)
Frédérique Tellier et al.
Pest management science, 65(2), 129-136 (2008-10-28)
The metabolism of cymoxanil [1-(2-cyano-2-methoxyiminoacetyl)-3-ethylurea] and fungicidal cyanooxime analogues was monitored on three phenotypes of Botrytis cinerea Pers. ex Fr. differing in their sensitivity towards cymoxanil. For this purpose, labelled [2-(14)C]cymoxanil was added either to the culture medium of these
Teratological examination of the experimental fungicide DPX 3217 50 WP in CFY rats.
L Várnagy et al.
Acta veterinaria Academiae Scientiarum Hungaricae, 28(2), 223-231 (1980-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service