Skip to Content
Merck
All Photos(1)

Key Documents

45089

Supelco

Elaidic acid

analytical standard

Synonym(s):

trans-9-Octadecenoic acid, trans-Oleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726543
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

semisynthetic

Quality Level

grade

analytical standard

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

288 °C/100 mmHg (lit.)

mp

42-44 °C (lit.)

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+

InChI key

ZQPPMHVWECSIRJ-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 177.5 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 45089 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U McCloy et al.
Journal of lipid research, 45(3), 474-485 (2003-12-18)
Altered use of different dietary fatty acids may contribute to several chronic diseases, including obesity, noninsulin-dependent diabetes mellitus, and cardiovascular disease. However, few comparative data are available to support this link, so the goal of the present study was to
G Andrei et al.
Antiviral research, 45(3), 157-167 (2000-04-20)
A fatty acid derivative of ganciclovir (GCV), elaidic acid ganciclovir (E-GCV), has been evaluated for its inhibitory activity against laboratory and clinical strains of herpes simplex type 1 (HSV-1) and type 2 (HSV-2), varicella-zoster virus (VZV) and human cytomegalovirus (HCMV)
Nima Sanadgol et al.
Clinical biochemistry, 43(12), 968-972 (2010-05-11)
Elaidic acid, the predominant trans-fatty acid in industrially hydrogenated oils, exists on high levels in Iranian hydrogenated oils and margarines. This study was undertaken to investigate the effect of elaidic acid and its cis-counterpart oleic acid on expression of ICAM-1
Véronique Chajès et al.
Nutrition and cancer, 63(8), 1235-1250 (2011-11-03)
Elaidic acid is the main unnatural trans fatty acid isomer occurring during partial hydrogenation of vegetable oils used as ingredients for the formulation of processed foods. The main objective is to assess associations between processed food intakes and plasma phospholipid
Patricia Burriel et al.
Physical review letters, 100(13), 134503-134503 (2008-06-04)
An anomalously long transient is needed to achieve a steady pressurization of a fluid when forced to flow through micronarrowed channels under constant mechanical driving. This phenomenon, known as the "bottleneck effect" is here revisited from a different perspective, by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service