Skip to Content
Merck
All Photos(2)

Key Documents

37887

Supelco

Butachlor

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26ClNO2
CAS Number:
Molecular Weight:
311.85
Beilstein:
2873811
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOCN(C(=O)CCl)c1c(CC)cccc1CC

InChI

1S/C17H26ClNO2/c1-4-7-11-21-13-19(16(20)12-18)17-14(5-2)9-8-10-15(17)6-3/h8-10H,4-7,11-13H2,1-3H3

InChI key

HKPHPIREJKHECO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wan-Yi Liu et al.
Ecotoxicology (London, England), 20(2), 377-384 (2011-01-07)
Butachlor is the most commonly used herbicide on paddy fields in Taiwan and throughout Southeast Asia. Since paddy fields provide habitat for pond breeding amphibians, we examined growth, development, time to metamorphosis, and survival of alpine cricket frog tadpoles (Fejervarya
S Dwivedi et al.
Letters in applied microbiology, 51(1), 54-60 (2010-05-19)
Isolation, characterization and assessment of butachlor-degrading potential of bacterial strain JS-1 in soil. Butachlor-degrading bacteria were isolated using enrichment culture technique. The morphological, biochemical and genetic characteristics based on 16S rDNA sequence homology and phylogenetic analysis confirmed the isolate as
Changming Yang et al.
Journal of environmental quality, 41(3), 716-723 (2012-05-09)
Biogeochemical processes in riparian zones regulate contaminant movement to receiving waters and often mitigate the impact of upland sources of contaminants on water quality. However, little research has been reported on the microbial process and degradation potential of herbicide in
Nidhi Kumari et al.
Chemosphere, 77(11), 1501-1507 (2009-11-03)
The present study examines butachlor-induced inhibition of growth, photosynthetic pigments such as chlorophyll a, phycocyanin, allophycocyanin, phycoerythrin, photosystems I and II, whole chain electron transport, oxygen evolution, carbon fixation, ATP content, total thiol and glutathione contents of Aulosira fertilissima. For
Hongzhi He et al.
Ecotoxicology and environmental safety, 80, 91-96 (2012-03-03)
Both single and joint toxicity of atrazine and butachlor to freshwater green alga Scenedesmus obliquus and cladoceran Daphnia carinata isolated from South China were investigated in the present study. The 96 h-EC(50) values of atrazine and butachlor to S. obliquus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service