Skip to Content
Merck
All Photos(2)

Key Documents

36163

Supelco

Metolachlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(-2-methoxy-1-methylethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Beilstein:
2743537
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.526-1.529

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cccc(C)c1N(C(C)COC)C(=O)CCl

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3

InChI key

WVQBLGZPHOPPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Metolachlor is a commonly used chloroacetamide[1][2]preemergent pesticide, which can be used for weed control management in a variety of food crops.[3] It can undergo degradation in soil via microbial transformation reactions and hydrolytic reactions.[3]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photocatalytic-assisted ozone degradation of metolachlor aqueous solution
Orge.A.C, et al.
Chemical Engineering Journal, 318, 247-253 (2017)
Metolachlor photocatalytic degradation using TiO 2 photocatalysts
Sakkas.A.V
Applied Catalysis. B, Environmental, 49(3), 195-205 (2004)
Metolachlor photodegradation study in aqueous media under natural and simulated solar irradiation
Dimou.DA, et al.
Journal of Agricultural and Food Chemistry, 53(3), 694- 701 (2005)
J Restivo et al.
Journal of hazardous materials, 239-240, 249-256 (2012-09-27)
The catalytic ozonation of the herbicide metolachlor (MTLC) was tested using carbon nanomaterials as catalysts. Multiwalled carbon nanotubes were used in semi-batch experiments and carbon nanofibres grown on a honeycomb cordierite monolith were tested in continuous experiments. The application of
Richard E Lizotte et al.
The Science of the total environment, 427-428, 373-381 (2012-05-09)
We examined the mitigation efficiency of a managed riverine wetland amended with a mixture of suspended sediment, two nutrients (nitrogen and phosphorus), and three pesticides (atrazine, metolachlor, and permethrin) during a simulated agricultural runoff event. Hydrologic management of the 500

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service