Skip to Content
Merck
All Photos(1)

Key Documents

36927

Supelco

Benzyl butyl phthalate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[CH3(CH2)3O2C]C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
312.36
Beilstein:
2062204
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

10.8 (vs air)

vapor pressure

0.16 mmHg ( 150 °C)

autoignition temp.

450 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.54 (lit.)

density

1.1 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCc2ccccc2

InChI

1S/C19H20O4/c1-2-3-13-22-18(20)16-11-7-8-12-17(16)19(21)23-14-15-9-5-4-6-10-15/h4-12H,2-3,13-14H2,1H3

InChI key

IRIAEXORFWYRCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Ching Chen et al.
Ecotoxicology and environmental safety, 74(5), 1325-1330 (2011-04-19)
This study examined the effect of benzyl butyl phthalate (BBP), a phthalate ester (PAE) and an endocrine disruptor, on water celery, Ipomoea aquatica Forsk., one of the most popular leaf vegetables in Taiwan. After 28 days of cultivation, treatment with
Jan L Lyche et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 12(4), 225-249 (2010-02-26)
The purposes of this review are to (1) evaluate human and experimental evidence for adverse effects on reproduction and development in humans, produced by exposure to phthalates, and (2) identify knowledge gaps as for future studies. The widespread use of
Allan C Just et al.
Environmental health perspectives, 120(10), 1475-1480 (2012-06-27)
Recent cross-sectional studies suggest a link between butylbenzyl phthalate (BBzP) in house dust and childhood eczema. We aimed to evaluate whether concentrations of monobenzyl phthalate (MBzP), the main BBzP metabolite in urine, during pregnancy are associated prospectively with eczema in
Raquel Moral et al.
Environmental health : a global access science source, 10(1), 5-5 (2011-01-19)
Environmental estrogens are exogenous estrogen-mimicking compounds that can interfere with endogenous endocrine systems. Several of these endocrine disruptors have been shown to alter normal development and influence tumorigenesis in experimental models. N-butyl benzyl phthalate (BBP), a widely used plasticizer, is
José-Luis Martínez-Guitarte et al.
Aquatic toxicology (Amsterdam, Netherlands), 110-111, 84-90 (2012-01-27)
Non-coding RNAs (ncRNAs) represent an important transcriptional output of eukaryotic genomes. In addition to their functional relevance as housekeeping and regulatory elements, recent studies have suggested their involvement in rather unexpected cellular functions. The aim of this work was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service