Skip to Content
Merck
All Photos(1)

Documents

152641

Sigma-Aldrich

Diisobutyl phthalate

99%

Synonym(s):

DIBP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2CH2CH(CH3)2]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
2054802
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.49 (lit.)

bp

327 °C (lit.)

density

1.039 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C

InChI

1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3

InChI key

MGWAVDBGNNKXQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Effect of an Environmentally Relevant Phthalate Mixture on Primordial Germ Cells of Xenopus laevis Embryos.
Nathalie Momplaisir et al.
microPublication biology, 2018 (2018-12-03)
Feng Zeng et al.
Journal of hazardous materials, 164(2-3), 1171-1178 (2008-10-31)
Surface soils from 37 sampling sites including roadsides (RS), parks (PA) and residential areas (RE) of the Subtropical City, Guangzhou, were collected in December of 2005 and analyzed for 16 phthalate esters (PAEs). PAEs were detected in all surface soils
Julie Borch et al.
Toxicology letters, 163(3), 183-190 (2006-02-07)
Phthalates are widely used as plasticizers in various consumer products and building materials. Some of the phthalates are known to interfere with male reproductive development in rats, and di-n-butyl phthalate (DBP), diethylhexyl phthalate (DEHP) and butyl benzyl phthalate (BBP) were
Julie Boberg et al.
Toxicology, 250(2-3), 75-81 (2008-07-08)
Endocrine disrupting chemicals can induce malformations and impairment of reproductive function in experimental animals and may have similar effects in humans. Recently, the environmental obesogen hypothesis was proposed, suggesting that environmental chemicals contribute to the development of obesity and insulin
Kyunghee Ji et al.
Environmental research, 110(4), 375-382 (2010-03-17)
Diet is purported to be means of exposure to many environmental contaminants. The purpose of this study is to understand the influence of dietary change on the levels of exposure to several environmental chemicals - in particular, antibiotics and phthalates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service