Skip to Content
Merck
All Photos(1)

Key Documents

36124

Supelco

Chlorfenson

PESTANAL®, analytical standard

Synonym(s):

4-Chlorophenyl 4-chlorobenzenesulfonate, Difenson, Ovex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2O3S
CAS Number:
Molecular Weight:
303.16
Beilstein:
2944674
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(OS(=O)(=O)c2ccc(Cl)cc2)cc1

InChI

1S/C12H8Cl2O3S/c13-9-1-5-11(6-2-9)17-18(15,16)12-7-3-10(14)4-8-12/h1-8H

InChI key

RZXLPPRPEOUENN-UHFFFAOYSA-N

General description

Chlorfenson is a pyrethroid pesticide effective against mites of citrus fruit, vegetable crops and ornamentals. It is a non-systemic acaricide with residual ovicidal activity.

Application

Chlorfenson may be used as a reference standard for the determination of chlorfenson in bee pollens by using a solid-phase extraction (SPE) column for cleanup followed by gas chromatography using electron-capture detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE DANGER OF USING PESTICIDES TO CONTROL VARROA JACOBSONI OUD.
Arzone A
Apiacta (1984)
Simultaneous determination of organochlorine, organophosphorus, and pyrethroid pesticides in bee pollens by solid-phase extraction cleanup followed by gas chromatography using electron-capture detector.
Zhang L, et al.
Food Analytical Methods, 6(6), 1508-1514 (2013)
Gustavo A Alonso et al.
Sensors (Basel, Switzerland), 11(4), 3791-3802 (2011-12-14)
An approach to an inhibition bioelectronic tongue is presented. The work is focused on development of an automated flow system to carry out experimental assays, a custom potentiostat to measure the response from an enzymatic biosensor, and an inhibition protocol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service