Skip to Content
Merck
All Photos(2)

Key Documents

45376

Supelco

Chlorobenzilate

PESTANAL®, analytical standard

Synonym(s):

Ethyl 4,4′-dichlorobenzilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14Cl2O3
CAS Number:
Molecular Weight:
325.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C(O)(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C16H14Cl2O3/c1-2-21-15(19)16(20,11-3-7-13(17)8-4-11)12-5-9-14(18)10-6-12/h3-10,20H,2H2,1H3

InChI key

RAPBNVDSDCTNRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carcinogenicity of Chlorobenzilate in mice, rats, and dogs.
M D Reuber
Clinical toxicology, 16(1), 67-98 (1980-03-01)
Urinary chlorobenzilate residues in citrus fieldworkers.
J Griffith et al.
Bulletin of environmental contamination and toxicology, 35(4), 496-499 (1985-10-01)
Chlorobenzilate residues in citrus-worker urine.
K A Levy et al.
Bulletin of environmental contamination and toxicology, 27(2), 235-238 (1981-08-01)
Designing safer (soft) drugs by avoiding the formation of toxic and oxidative metabolites.
Nicholas Bodor et al.
Methods in molecular biology (Clifton, N.J.), 186, 301-312 (2002-05-17)
G M Wang
Regulatory toxicology and pharmacology : RTP, 4(4), 361-371 (1984-12-01)
In reaching a regulatory decision on the use of pesticides with carcinogenic potential, it is of great importance to investigate the extent of dermal exposure and absorption of a pesticide to users and field workers. By applying this information, along

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service