Skip to Content
Merck
All Photos(1)

Key Documents

33674

Supelco

N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine

PESTANAL®, analytical standard

Synonym(s):

Acetamiprid, sum of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClN4
CAS Number:
Molecular Weight:
222.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

100-102 °C

application(s)

agriculture
environmental

format

neat

SMILES string

CN(Cc1ccc(Cl)nc1)\C(C)=N\C#N

InChI

1S/C10H11ClN4/c1-8(14-7-12)15(2)6-9-3-4-10(11)13-5-9/h3-5H,6H2,1-2H3/b14-8+

InChI key

WCXDHFDTOYPNIE-RIYZIHGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetamiprid, is a neonicotinoid insecticide, which finds applications in agriculture for pest control. It can also act as acetylcholine nicotinic receptor.
Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Madianos et al.
Biosensors & bioelectronics, 101, 268-274 (2017-11-03)
A novel impedimetric biosensor was developed for the detection of the two extensively used pesticides, acetamiprid and atrazine. By employing the sputtering and e-beam lithography techniques, platinum nanoparticles (Pt NPs) were deposited in a bridge-like arrangement, in between interdigitated electrodes
Comparative toxicity of foliar and systemic applications of acetamiprid and imidacloprid against the cotton whitefly, Bemisia tabaci (Hemiptera: Aleyrodidae)
Horowitz .R.A, et al.
Bulletin of Entomological Research, 88, 437-442 (1998)
Analysis of acetamiprid in vegetables using gas chromatography-tandem mass spectrometry
Mateu-Sanchez M, et al.
Analytical Sciences, 19, 701-704 (2003)
Theofylaktos Apostolou et al.
Biosensors, 10(2) (2020-01-30)
Population growth and increased production demands on fruit and vegetables have driven agricultural production to new heights. Nevertheless, agriculture remains one of the least optimized industries, with laboratory tests that take days to provide a clear result on the chemical
Seyed Mohammad Taghdisi et al.
Food chemistry, 268, 342-346 (2018-08-02)
As one of the most toxic mycotoxins, aflatoxin B1 (AFB1) is a major food pollutant which can pose a high risk to human health. In this work, an accurate fluorescent sensing method was proposed for AFB1 determination, based on hairpin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service