Skip to Content
Merck
All Photos(1)

Documents

34227

Supelco

Imazamox

PESTANAL®, analytical standard

Synonym(s):

2-(4-Isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methoxymethylnicotinic acid, 2-[4,5-Dihydro-4-methyl-4-(1-methylethyl)-5-oxo-1H-imidazol-2-yl]-5-(methoxymethyl)-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N3O4
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COCc1cnc(C2=NC(C)(C(C)C)C(=O)N2)c(c1)C(O)=O

InChI

1S/C15H19N3O4/c1-8(2)15(3)14(21)17-12(18-15)11-10(13(19)20)5-9(6-16-11)7-22-4/h5-6,8H,7H2,1-4H3,(H,19,20)(H,17,18,21)

InChI key

NUPJIGQFXCQJBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazamox is an imidazolinone herbicide which acts by inhibiting acetohydroxyacid synthase activity.

Application

Imazamox may be used as an analytical reference standard for the determination of the analyte in environmental water samples, soil and in leaves of wheat plants by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alterations induced by imazamox on acetohydroxyacid synthase activity of common bean (Phaseolus vulgaris) depend on leaf position.
Garcia-Garijo A, et al.
Pesticide Biochemistry and Physiology, 104(1), 72-76 (2012)
E Kathryn Barto et al.
PloS one, 6(11), e27195-e27195 (2011-11-24)
Allelopathy, a phenomenon where compounds produced by one plant limit the growth of surrounding plants, is a controversially discussed factor in plant-plant interactions with great significance for plant community structure. Common mycorrhizal networks (CMNs) form belowground networks that interconnect multiple
Quantitative analysis of imazamox herbicide in environmental water samples by capillary electrophoresis electrospray ionization mass spectrometry.
Safarpour H, et al.
Journal of Chromatography A, 1036(2), 217-222 (2004)
Ultrasound-assisted Extraction with LC-TOF/MS Identification and LC-UV Determination of Imazamox and its Metabolites in Leaves of Wheat Plants.
Rojano-Delgado AM, et al.
Phytochemical Analysis, 25(4), 357-363 (2014)
A simple and efficient method for imidazolinone herbicides determination in soil by ultra-high performance liquid chromatography-tandem mass spectrometry.
Kemmerich M, et al.
Journal of Chromatography A, 1412(2), 82-89 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service