Skip to Content
Merck
All Photos(1)

Key Documents

45555

Supelco

MCPA

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-2-methylphenoxyacetic acid, 4-Chloro-o-tolyloxyacetic acid, MCPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)OCH2CO2H
CAS Number:
Molecular Weight:
200.62
Beilstein:
2051752
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

114-118 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(Cl)ccc1OCC(O)=O

InChI

1S/C9H9ClO3/c1-6-4-7(10)2-3-8(6)13-5-9(11)12/h2-4H,5H2,1H3,(H,11,12)

InChI key

WHKUVVPPKQRRBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Wirsching et al.
Frontiers in microbiology, 11, 2107-2107 (2020-09-29)
The objective of our study was to test whether limited microbial degradation at low pesticide concentrations could explain the discrepancy between overall degradability demonstrated in laboratory tests and their actual persistence in the environment. Studies on pesticide degradation are often
G G Bond et al.
British journal of industrial medicine, 50(4), 340-348 (1993-04-01)
For the purpose of assessing the human carcinogenic potential of the chlorophenoxy herbicides MCPA, MCPP, and 2,4-DP, the relevant epidemiological and toxicological evidence is reviewed. These compounds have not produced tumours in animal studies conducted under current test guidelines, giving
Nora Badawi et al.
Environmental pollution (Barking, Essex : 1987), 170, 32-38 (2012-07-06)
Soil suspensions (slurries) are commonly used to estimate the potential of soil microbial communities to mineralize organic contaminants. The preparation of soil slurries disrupts soil structure, however, potentially affecting both the bacterial populations and their protozoan predators. We studied the
Meriç Batıoğlu-Pazarbaşı et al.
FEMS microbiology ecology, 80(2), 331-341 (2012-05-23)
Centimetre-scale vertical distribution of mineralization potential was determined for 2,4-dichlorophenoxyacetic acid (2,4-D), 4-chloro-2-methylphenoxyacetic acid (MCPA) and 2-(4-chloro-2-methylphenoxy)propanoic acid (MCPP) by 96-well microplate radiorespirometric analysis in aquifer sediment sampled just below the groundwater table. Mineralization of 2,4-D and MCPA was fastest
Ali Yuzir et al.
Bioresource technology, 109, 31-37 (2012-02-10)
The degradation of (RS)-MCPP was investigated in an anaerobic membrane bioreactor (AnMBR) using nitrate as an available electron acceptor under different COD/NO(3)(-)-N ratios. Results showed high soluble COD removal efficiency (80-93%) when the reactor was operated at high COD/NO(3)(-)-N ratios.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service