Skip to Content
Merck
All Photos(1)

Documents

33738

Supelco

Baythroid®

PESTANAL®, analytical standard

Synonym(s):

Cyfluthrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18Cl2FNO3
CAS Number:
Molecular Weight:
434.29
Beilstein:
2788149
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2ccc(F)c(Oc3ccccc3)c2

InChI

1S/C22H18Cl2FNO3/c1-22(2)15(11-19(23)24)20(22)21(27)29-18(12-26)13-8-9-16(25)17(10-13)28-14-6-4-3-5-7-14/h3-11,15,18,20H,1-2H3

InChI key

QQODLKZGRKWIFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Baythroid is an insecticide, that can be effectively used to control sucking insect pests in cotton crops like jassids, whiteflies and also thrips, which can suck out juices from these plants and hence, damage them.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Baythroid is a registered trademark of Bayer AG
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farmers' Bulletin (1975)
Christophe Antonio-Nkondjio et al.
Parasites & vectors, 6(1), 10-10 (2013-01-15)
Insecticide treated materials remain the mainstay for malaria prevention. The current study reports on the entomological impact of cyfluthrin impregnated bed nets on malaria transmission in Mbandjock, a semi urban locality in southern Cameroon. Several findings pertaining to the recent
Shaohua Chen et al.
Bioresource technology, 132, 16-23 (2013-02-12)
Brevibacterium aureum DG-12, a new bacterial strain isolated from active sludge, was able to degrade and utilize cyfluthrin as a growth substrate in the mineral medium. Response surface methodology using central composite rotatable design of cultural conditions was successfully employed
Changlu Wang et al.
Journal of economic entomology, 106(1), 347-352 (2013-03-02)
Current bed bug, Cimex lectularius L., control usually involves insecticide applications that pose a high risk of insecticide exposure to residents and applicators. To minimize these risks and the amount of insecticides used, we designed and evaluated a dust band
Christos G Athanassiou et al.
Journal of economic entomology, 106(2), 1064-1070 (2013-06-22)
The insecticidal effect of Temprid, a formulation that contains beta-cyfluthrin and imidacloprid, was tested on concrete for control of seven stored-product insect species: the rusty grain beetle, Cryptolestes ferrugineus (Stephens); the sawtoothed grain beetle, Oryzaephilus surinamensis (L.); the red flour

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service