Skip to Content
Merck
All Photos(2)

Documents

33396

Supelco

Allethrin

PESTANAL®, analytical standard

Synonym(s):

Allylcinerine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Beilstein:
2294836
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C/C1C(C(=O)OC2CC(=O)C(CC=C)=C2C)C1(C)C

InChI

1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3

InChI key

ZCVAOQKBXKSDMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Allethrin is a non-cyano containing pyrethroid household insecticide, that can be used to control flies and mosquitoes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Acton A.Q et al.
Issues in Genetic Research (2013)
Singh.KD
Pesticide Chemistry and Toxicology Volume 1 of Toxicology: Agriculture and Environment (2012)
Hitoshi Kawada et al.
PloS one, 6(8), e22574-e22574 (2011-08-20)
Anopheles gambiae s.s., Anopheles arabiensis, and Anopheles funestus s.s. are the most important species for malaria transmission. Pyrethroid resistance of these vector mosquitoes is one of the main obstacles against effective vector control. The objective of the present study was
Vadoud H Niri et al.
Journal of separation science, 32(7), 1075-1080 (2009-03-07)
The possibility of sampling the free and particle-bound concentrations of organic compounds was studied using two different sampling techniques at the same time: needle trap device (NTD) and solid-phase microextraction (SPME). In this study, a mosquito coil was used to
April P Neal et al.
Toxicological sciences : an official journal of the Society of Toxicology, 116(2), 604-613 (2010-05-15)
Pyrethroid insecticides are one of the most widely used classes of insecticides. Previous studies revealed that pyrethroids potently affect the insect voltage-gated sodium (Na(+)) channel (VGSC), resulting in prolonged channel open time. However, recent findings have suggested that pyrethroids may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service