Skip to Content
Merck
All Photos(1)

Documents

33506

Sigma-Aldrich

Oxalic acid dihydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (manganometric)

Synonym(s):

Ethanedioic acid dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCO2H · 2H2O
CAS Number:
Molecular Weight:
126.07
Beilstein:
3679436
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021701
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

4.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99.5% (manganometric)

form

crystals

impurities

foreign organic matter, in accordance
≤0.001% total nitrogen (N)

ign. residue

≤0.01% (as SO4)

mp

104-106 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[H]O[H].[H]O[H].OC(=O)C(O)=O

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxalic acid dihydrate crystals are monoclinic and exhibit a space group P21/n. Each unit cell contains two molecules. 13C chemical shielding tensors in oxalic acid dihydrate have been evaluated.

Application

Oxalic acid dihydrate has been used to prepare oxalate precursor, which was employed for the synthesis of zinc oxide nanopowders. It may be used to prepare the 0.01M solution of oxalic acid, which was employed in the mobile phase in liquid chromatographic studies.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jungang Jiang et al.
Carbohydrate polymers, 247, 116727-116727 (2020-08-25)
Nanocellulose is a promising material but its isolation generally requires unrecyclable hazardous chemicals and high energy consumption and its overall yield is low due to the use of high purity cellulose as precursor. In order to overcome these shortcomings, in
José Miguel Hidalgo Herrador et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-23)
The use of renewable local raw materials to produce fuels is an important step toward optimal environmentally friendly energy consumption. In addition, the use of these sources together with fossil fuels paves the way to an easier transition from fossil
The effect of the rate of precursor production on the purity and aggregation morphology of nanoparticulate zinc oxide.
Duffy GM, et al.
Journal of Materials Chemistry, 17(2), 181-184 (2007)
13C chemical shielding in oxalic acid, oxalic acid dihydrate, and diammonium oxalate.
Griffin RG, et al.
J. Chem. Phys. , 63(3), 1267-1271 (1975)
Francesca Capolongo et al.
Journal of AOAC International, 85(1), 8-14 (2002-03-07)
Residues of oxytetracycline (OTC) in edible tissues (muscle, liver, and kidney) of 18 turkeys were determined after continuous administration of the drug for 3 days in drinking water at the maximum recommended concentration of 400 mg/L. The European Union (EU)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service