Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Oxalic acid dihydrate
Synonym(s):
Oxalic acid dihydrate, Ethanedioic acid dihydrate
Linear Formula:
HO
2
CCO
2
H · 2H
2
O
CAS Number:
6153-56-6
Molecular Weight:
126.07
EC Number:
205-634-3
Products
Product Category
acids (5)
organic acids (3)
building blocks (2)
Brand
Sigma-Aldrich (7)
Supelco (4)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
1.00495
Oxalic acid dihydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
8.18242
Oxalic acid dihydrate,
for synthesis
Expand
View Pricing
247537
Oxalic acid dihydrate,
ACS reagent, ≥99%
Expand
View Pricing
1.00489
Oxalic acid dihydrate,
Suprapur
®
Expand
View Pricing
1.00492
Oxalic acid dihydrate,
EMPLURA
®
Expand
View Pricing
75699
Oxalic acid dihydrate,
BioUltra, ≥99.0% (RT)
Expand
View Pricing
33506
Oxalic acid dihydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (manganometric)
Expand
View Pricing
1.93418
Oxalic acid dihydrate,
EMPLURA
®
Expand
View Pricing
1.93018
Oxalic acid dihydrate,
for analysis EMPARTA
®
ACS
Expand
View Pricing
O0376
Oxalic acid dihydrate,
ReagentPlus
®
, ≥99.0% (GC)
Expand
View Pricing
23-0820
Oxalic acid dihydrate,
JIS special grade, 99.5-100.2%
Expand
View Pricing
23-0830
Oxalic acid dihydrate,
SAJ first grade, 99.0-101.0%
Expand
View Pricing