MX1390
NMP
OmniSolv®
Synonym(s):
1-Methyl-2-Pyrrolidone, N-Methyl-2-Pyrrolidinone,NMP, Methyl Pyrrolidone
About This Item
LC/MS: suitable
gas chromatography (GC): suitable
Recommended Products
Product Name
1-Methyl-2-pyrrolidone, OmniSolv®
vapor pressure
0.32 hPa ( 20 °C)
Quality Level
product line
OmniSolv®
Assay
≥99.7% (GC)
form
liquid
technique(s)
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
impurities
≤0.08% Water (H2O)
≤1 μeq/g Titrable base
ign. residue
≤5 ppm
color
APHA: ≤10
pH
8.5-10.0 (20 °C, 100 g/L in H2O)
bp
202 °C/1013 hPa
mp
-24 °C
transition temp
flash point 91 °C
solubility
1000 g/L
density
1.03 g/cm3 at 20 °C
UV cutoff
≤270 nm
UV absorption
λ: 270 nm Amax: ≤1.00
λ: 300 nm Amax: ≤0.20
λ: 325 nm Amax: ≤0.10
λ: 350 nm Amax: ≤0.03
λ: 400 nm Amax: ≤0.005
shipped in
ambient
storage temp.
room temp
SMILES string
N1(CCCC1=O)C
InChI
1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3
InChI key
SECXISVLQFMRJM-UHFFFAOYSA-N
Related Categories
Application
Preparation Note
Analysis Note
Legal Information
greener alternative product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
195.8 °F - Pensky-Martens closed cup
Flash Point(C)
91 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service