Skip to Content
Merck
All Photos(1)

Key Documents

31558

Supelco

Malathion solution

100 μg/mL in cyclohexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O6PS2
CAS Number:
Molecular Weight:
330.36
Beilstein:
1804525
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC

InChI

1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3

InChI key

JXSJBGJIGXNWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malathion is an organophosphate mostly used as pesticide.

Application

Malathion may be used as standard in determining Malathion in pesticide emulsifiable concentrate formulations using Fourier transform–Raman (FT–Raman) spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

FT?Raman spectrometry determination of Malathion in pesticide formulations.
Quintas, Guillermo, Salvador Garrigues, and Miguel de la Guardia.
Talanta, 63.2 , 345-350 (2004)
Oxidative stress and cholinesterase inhibition in saliva and plasma of rats following subchronic exposure to malathion.
Abdollahi M
Comparative Biochemistry and Physiology. Toxicology & Pharmacology : CBP, 137(1), 29-34 (2004)
David Jira et al.
Neuro endocrinology letters, 33 Suppl 3, 53-59 (2013-01-29)
Malathion is generally not classified as toxic. However, the toxicity seems to be species-dependent. Local and systemic toxicity data for birds are rare, but a decrease of wild bird densities in areas where malathion was applied was reported. Aim of
S Abdul Majeed et al.
Cell biology and toxicology, 29(1), 59-73 (2012-12-12)
Rohu gill cell line (LRG) was established from gill tissue of Indian major carp (Labeo rohita), a freshwater fish cultivated in India. The cell line was maintained in Leibovitz's L-15 supplemented with 10 % foetal bovine serum (FBS). This cell line
Lorin A Neuman-Lee et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(6), 442-448 (2013-03-05)
This study examined the effects of sub-lethal exposure of the ubiquitous pesticide malathion on the behavior of the model orthopteran species, the house cricket (Acheta domesticus). Increasing concentrations of malathion caused male crickets to increase periods of non-directional movement, such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service