Merck
  • Sublethal contaminant exposure alters behavior in a common insect: important implications for trophic transfer.
Sublethal contaminant exposure alters behavior in a common insect: important implications for trophic transfer.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2013-03-05)
Lorin A Neuman-Lee, Gareth R Hopkins, Edmund D Brodie, Susannah S French
ABSTRACT
This study examined the effects of sub-lethal exposure of the ubiquitous pesticide malathion on the behavior of the model orthopteran species, the house cricket (Acheta domesticus). Increasing concentrations of malathion caused male crickets to increase periods of non-directional movement, such as twitching and grooming, directional movement, and to seek out shelter less. These are all behavioral alterations that may increase the cricket's chances of being preyed upon, and thus have the potential for serious ecological consequences through trophic transfer. This study also revealed that female crickets appeared to be less affected by malathion than their male conspecifics, indicating a potential sex-bias in both susceptibility and possible predator attack.
Product Description
Supelco
Malathion, certified reference material, TraceCERT®
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Malathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard