Skip to Content
Merck
All Photos(1)

Key Documents

BX1795

Supelco

sec-Butyl Alcohol

Synonym(s):

sec-Butyl Alcohol, 2-Butanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH3
CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-158-5
Assay:
≥98% (GC)
bp:
99-102 °C/1013 hPa
vapor pressure:
16.5 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

16.5 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

color

APHA: ≤20

pH

7 (20 °C)

bp

99-102 °C/1013 hPa

mp

-115 °C

transition temp

flash point 24 °C

solubility

125 g/L

density

0.81 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OC(CC)C

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3

InChI key

BTANRVKWQNVYAZ-UHFFFAOYSA-N

Analysis Note

Assay (GC): 98.0% min
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service