Skip to Content
Merck
All Photos(1)

Key Documents

82303

Millipore

Casein Peptone

Enzymatic digest of casein from bovine milk, suitable for biotechnology and microbiology

Synonym(s):

Peptone from casein, Peptone from casein, enzymatic digest

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41106212
eCl@ss:
42040102
NACRES:
NA.85

product name

Peptone from casein, enzymatic digest, Enzymatic digest of casein, for biotechnological purposes

biological source

bovine milk

Quality Level

grade

for biotechnological purposes

form

powder

composition

total nitrogen (N), ~13%

nitrogen analysis

, ~0.3 AN:TN ratio

ign. residue

<17%

loss

<6% loss on drying

pH

7.0±0.5 (2% in H2O)

solubility

H2O: 2%, clear, yellow to tan
water: soluble 2%, clear, faintly brown-yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone from an enzymatic digest of casein is used as a nitrogen source in culture media formulated for isolating and cultivating fastidious and non-fastidious bacteria and fungi. It is a refined hydrolysate with high solubility and is used as a microbiological nutrient. It is a rich source of amino acids and peptides recommended for culture media in analytical microbiology and industrial fermentation.

Application

Peptone from casein may be used as medium supplement in the following:
Bacterial cultures for short term storage of bacteria.
The basal Glucose Yeast Peptone (GYP) medium.
As nitrogen supplement in the culture medium of Ceriporiopsis subvermispora.
Culture medium of Methylobacterium rhodesianum and Ralstonia eutropha.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Kai et al.
Archives of microbiology, 187(5), 351-360 (2006-12-21)
Bacterial antagonists are bacteria that negatively affect the growth of other organisms. Many antagonists inhibit the growth of fungi by various mechanisms, e.g., secretion of lytic enzymes, siderophores and antibiotics. Such inhibition of fungal growth may indirectly support plant growth.
The production of polyhydroxybutyrate by Methylobacterium rhodesianum and Ralstonia eutropha in media containing glycerol and casein hydrolysates.
Bormann EJ and Roth M.
Biotechnology Letters, 21(12), 1059-1063 (1999)
Wolfgang Harreither et al.
Applied and environmental microbiology, 75(9), 2750-2757 (2009-03-10)
Cellobiose dehydrogenase (CDH), an extracellular flavocytochrome produced by several wood-degrading fungi, was detected in cultures of the selective delignifier Ceriporiopsis subvermispora when grown on a cellulose- and yeast extract-based liquid medium. CDH amounted to up to 2.5% of total extracellular
Christiane Galhaup et al.
Microbiology (Reading, England), 148(Pt 7), 2159-2169 (2002-07-09)
The major laccase isoenzyme LAP2 secreted by the white-rot basidiomycete Trametes pubescens in response to high copper concentrations was purified to apparent electrophoretic homogeneity using anion-exchange chromatography and gel filtration. The monomeric protein has a molecular mass of 65 kDa
Holly Newton et al.
Nature communications, 11(1), 4653-4653 (2020-09-18)
Cancer cells demand excess nutrients to support their proliferation, but how tumours exploit extracellular amino acids during systemic metabolic perturbations remain incompletely understood. Here, we use a Drosophila model of high-sugar diet (HSD)-enhanced tumourigenesis to uncover a systemic host-tumour metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service