Skip to Content
Merck
All Photos(1)

Key Documents

181676

Supelco

Poly(Bisphenol A carbonate)

analytical standard, molecular weight series

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, molecular weight series

Quality Level

mol wt

average Mn 12,000-23,000 (typical)
average Mw 22,000-40,000 (typical)

technique(s)

gel permeation chromatography (GPC): suitable

density

1.2 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.CH2O3/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;2-1(3)4/h3-10,16-17H,1-2H3;(H2,2,3,4)

InChI key

XSXWYGABGYBZRM-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property. It can be synthesized by inducing supercritical CO2 to crystallinity in low molecular weight polycarbonate beads using solid-state polymerization method.

Application

It was used to study the lipase degradation and monitored using gel permeation chromatography (GPC).

Analysis Note

A lot specific analysis is included with each unit

Kit Components Only

Product No.
Description

  • average Mw 23,000; average Mn 13,000 25 g

  • average Mw 40,000; average Mn 21,000 25 g

  • average Mw 31,000; average Mn 18,000 25 g

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Crystallization and solid-state polymerization of poly (bisphenol A carbonate) facilitated by supercritical CO2.
Gross, Stephen M., et al.
Macromolecules, 33.1, 40-45 (2000)
Dynamics of lipase catalyzed enzymatic degradation of poly (bisphenol-A carbonate).
Sivalingam, G., and Giridhar Madras
Journal of Applied Polymer Science, 91.4, 2391-2396 (2004)
Leon Coulier et al.
Journal of chromatography. A, 1130(1), 34-42 (2006-06-01)
Hyphenation of infrared spectroscopy (IR) to liquid chromatography (LC) has been applied to study chemical changes in poly(bisphenol A)carbonate (PC) as a result of degradation. Especially coupling of LC to FTIR through solvent elimination is a sensitive approach to identify
M Yalon et al.
Journal of cataract and refractive surgery, 14(4), 393-395 (1988-07-01)
Bisphenol-A polycarbonate has been investigated as an improved polymer for ocular implants, especially for intraocular lenses (IOLs). Polycarbonate properties afford special opportunities for development of tougher, stronger, one-piece IOLs. Autoclave or gamma sterilizability and a higher refractive index may provide
M Utz et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 128(2), 217-227 (1997-11-14)
A method is proposed for the quantitative measurement of orientational anisotropy in glassy solids based on 2D dipolar NMR spectra with sample flipping (dipolar DECODER experiment). Purely dipolar spectra are obtained by chemical shift refocusing by a multiple pulse sequence.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service