Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(Bisphenol A carbonate)
CAS Number:
25037-45-0
Products
Brand
Sigma-Aldrich (2)
Supelco (2)
Gene Alias
Physical Form
Format
Quality Segment
Polymer Type
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
181625
Poly(Bisphenol A carbonate),
average M
w
~45,000 by GPC
Expand
View Pricing
435139
Poly(Bisphenol A carbonate),
melt index 10-12 g/10 min (300°C/1.2kg)
Expand
View Pricing
181641
Poly(Bisphenol A carbonate),
analytical standard
Expand
View Pricing
181676
Poly(Bisphenol A carbonate),
Expand
View Pricing