Skip to Content
Merck
All Photos(1)

Documents

17123

Millipore

Elliker Broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Lactobacillus Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

ascorbic acid, 0.5 g/L
casein enzymic hydrolysate, 20 g/L
dextrose, 5 g/L
gelatine, 2.5 g/L
lactose, 5 g/L
saccharose, 5 g/L
sodiumacetate, 1.5 g/L
sodium chloride, 4 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective for Lactobacillus spp.
nonselective for Lactococcus spp.
nonselective for Streptococcus spp.

Application

For culturing Streptococci and Lactobacilli of importance in the dairy industry.

Preparation Note

Suspend 48.5 g in 1 litre distilled water. Boil to dissolve the medium completely. Dispense as desired. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service