Skip to Content
Merck
All Photos(2)

Documents

10850

Millipore

D-(−)-Arabinose

suitable for microbiology, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723079
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

form

powder

optical activity

[α]20/D −104±2.0°, 24 hr, c = 10% in H2O

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

mp

162-164 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤25 mg/kg
Fe: ≤17 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤15 mg/kg

application(s)

microbiology

SMILES string

O[C@@H]1COC(O)[C@@H](O)[C@@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3-,4+,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-ZRMNMSDTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Arabinose is a rare aldopentose, and is rarely utilized by enteric bacteria as a source of carbon and energy. It is also found in the aloins of the plant genus Aloe and as a constituent of the polysaccharide of the bacterial genus Mycobacterium. Some of the enteric bacteria like Escherichia coli K-12 can mutate to utilize D-arabinose.

Application

D-(-)-Arabinose has been used as an inducer of λ-RED recombinant gene expression.

Biochem/physiol Actions

D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering complex biological systems in bacteria through recombinase-assisted genome engineering.
Santos CN and Yoshikuni Y2
Nature Protocols, 9, 1320-1336 (2014)
The Evolution of Metabolic Function.
Mortlock RP.
Science, 16-17 (1992)
Dushyant Mishra et al.
PLoS biology, 16(8), e2005570-e2005570 (2018-08-08)
Animals employ various types of taste receptors to identify and discriminate between different nutritious food chemicals. These macronutrients are thought to fall into 3 major groups: carbohydrates/sugars, proteins/amino acids, and fats. Here, we report that Drosophila larvae exhibit a novel
Agustina Llanos et al.
Microbial cell factories, 18(1), 14-14 (2019-01-30)
Research on filamentous fungi emphasized the remarkable redundancy in genes encoding hydrolytic enzymes, the similarities but also the large differences in their expression, especially through the role of the XlnR/XYR1 transcriptional activator. The purpose of this study was to evaluate
Nobuhiro Yamagata et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(2), 578-583 (2014-12-31)
Drosophila melanogaster can acquire a stable appetitive olfactory memory when the presentation of a sugar reward and an odor are paired. However, the neuronal mechanisms by which a single training induces long-term memory are poorly understood. Here we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service