Skip to Content
Merck
All Photos(1)

Key Documents

10839

Sigma-Aldrich

L-(+)-Arabinose

BioUltra, ≥99.5% (sum of enantiomers, HPLC)

Synonym(s):

Aldehydo-L-arabinose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723085
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.5% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]20/D +104.0±2.0°, 24 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.1%

loss

≤0.1% loss on drying

color

white

pH

4-7 (25 °C, 1 M in H2O)

mp

155-159 °C
160-163 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.04

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4-,5+/m0/s1

InChI key

PYMYPHUHKUWMLA-VAYJURFESA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Arabinose is used as a substrate to identify, differentiate and characterize pentose sugar isomerase(s). L-Arabinose is used in the bioproduction of L-ribose.

Biochem/physiol Actions

L-Arabinose is the naturally occurring isomer and is a constituent of plant polysaccharides. Most bacteria contain an inducible arabinose operon that codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Djursun Karasartova et al.
Journal of clinical immunology, 37(4), 388-396 (2017-05-11)
Splenectomy is associated with increased risk of overwhelming post-splenectomy infections despite proper anti-pneumococcal vaccination. As most studies concentrated on vaccination-induced humoral immunity, the cellular immune responses triggered in splenectomized patients are not yet well studied. The present study aims to
Daniel Concepcion et al.
Biology open, 6(7), 1065-1073 (2017-06-14)
Tbx6 is a T-box transcription factor with multiple roles in embryonic development as evidenced by dramatic effects on mesoderm cell fate determination, left/right axis determination, and somite segmentation in mutant mice. The expression of Tbx6 is restricted to the primitive
Chang-Su Park et al.
Applied microbiology and biotechnology, 92(6), 1187-1196 (2011-06-22)
A putative N-acyl-D-glucosamine 2-epimerase from Caldicellulosiruptor saccharolyticus was cloned and expressed in Escherichia coli. The recombinant enzyme was identified as a cellobiose 2-epimerase by the analysis of the activity for substrates, acid-hydrolyzed products, and amino acid sequence. The cellobiose 2-epimerase
M Helanto et al.
Applied microbiology and biotechnology, 83(1), 77-83 (2009-01-20)
L-Ribose is a rare and expensive sugar that can be used as a precursor for the production of L-nucleoside analogues, which are used as antiviral drugs. In this work, we describe a novel way of producing L-ribose from the readily
Koen J T Venken et al.
Nucleic acids research, 36(18), e114-e114 (2008-08-05)
Studying gene function in the post-genome era requires methods to localize and inactivate proteins in a standardized fashion in model organisms. While genome-wide gene disruption and over-expression efforts are well on their way to vastly expand the repertoire of Drosophila

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service