Skip to Content
Merck
All Photos(2)

Key Documents

06556

Sigma-Aldrich

Phenyl formate

≥98.0% (GC)

Synonym(s):

Phenoxyformaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOC6H5
CAS Number:
Molecular Weight:
122.12
Beilstein:
507113
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.511

bp

62 °C/1 mmHg (lit.)

density

1.120 g/mL at 20 °C (lit.)

functional group

phenoxy

SMILES string

O=COc1ccccc1

InChI

1S/C7H6O2/c8-6-9-7-4-2-1-3-5-7/h1-6H

InChI key

GEOWCLRLLWTHDN-UHFFFAOYSA-N

Gene Information

human ... STS(412)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the formylation of amines[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.L. Yale
The Journal of Organic Chemistry, 36, 3238-3238 (1971)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service