Skip to Content
Merck
All Photos(2)

Key Documents

04287

Supelco

Chlornitrofen

PESTANAL®, analytical standard

Synonym(s):

1,3,5-Trichloro-2-(4-nitrophenoxy)benzene, 4-Nitrophenyl 2,4,6-trichlorophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl3N1O3
CAS Number:
Molecular Weight:
318.54
Beilstein:
1890443
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)c1ccc(Oc2c(Cl)cc(Cl)cc2Cl)cc1

InChI

1S/C12H6Cl3NO3/c13-7-5-10(14)12(11(15)6-7)19-9-3-1-8(2-4-9)16(17)18/h1-6H

InChI key

XQNAUQUKWRBODG-UHFFFAOYSA-N

General description

Chlornitrofen (Cnf) is a diphenyl-ether-derived pesticide belonging to the nitrophenyl chemical class of compounds. It is a pre-emergence herbicide used to control annual weeds and some perennials, including Eleocharis acicularis and Sagittaria pygmaea in rice fields. The mode of action of Cnf is by the inhibition of a vital enzyme, protoporphyrinogen oxidase (PPO).

Application

Chlornitrofen may be used as an analytical reference standard for the determination of the analyte in river water, air samples and tobacco by chromatography based techniques.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

C-NMR and H-NMR conform to structure

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Mutagenicity of herbicide chlornitrofen (CNP) and its reduction products].
S Kitamori et al.
Sangyo eiseigaku zasshi = Journal of occupational health, 37(2), 143-144 (1995-03-01)
M Yamamoto et al.
The Tohoku journal of experimental medicine, 155(4), 385-386 (1988-08-01)
Rec assay of agricultural chemicals, MCPA-E, MCPA, CNP and PCNB was conducted. It was found that MCPA-E and MCPA accelerated the growth-inhibition originally induced by Mitomycin C in the rec- strain.
B M Francis
Teratology, 41(4), 443-451 (1990-04-01)
To determine the role of number and position of chlorine substituents on the developmental toxicity of diphenyl ether analogs of nitrofen, we have evaluated one unchlorinated, three monochlorinated, and five dichlorinated-phenyl 4'-nitrophenyl ethers with respect to effects on liver weight
B M Francis
Toxicology, 40(3), 297-309 (1986-09-01)
The fetotoxicity of nitrofen and 5 analogous diphenyl ethers was compared in mice, using survival on postnatal day 5 to evaluate prenatal and perinatal toxicity. The effect of each chemical on the Harderian gland weight of surviving pups was also
S Kitamori
Fukuoka igaku zasshi = Hukuoka acta medica, 87(6), 142-150 (1996-06-01)
Mutagenicities of three diphenyl ether-derived herbicides, chlornitrofen (CNP), nitrofen and chlormethoxynil, and their amino derivatives were assayed by the Ames test using TA98, TA100 and YG strains, recently developed tester strains. As for CNP, mutagenicities of its biodegradation products were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service