Skip to Content
Merck
All Photos(1)

Key Documents

00446

Millipore

Universal Microbial Limit Test Membrane

white colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

membrane size

70 mm

Quality Level

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
cosmetics
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Bacillus spp.
Escherichia coli
Pseudomonas spp.
Salmonella spp.
Staphylococcus spp.
coliforms

General description

Universal Microbial Limit Test Membrane are substrate-carrying ID membranes used for rapid identification and differentiation of colonies through characteristic color reactions by placing on the agar surface post-incubation. The membranes contain chromogenic substrates such as ONPG, X-Gal, or X-Glu, and other substrates and indicators, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane. This indicates microbial enzymatic activity, which serves to identify the genus and species. This economical, confirmatory, and rapid identification method is suitable for microorganisms from water, food, environmental, and clinical samples.

Application

Universal Microbial Limit Test Membrane is recommended for the detection of pathogenic microorganisms such as E. coli, S. aureus, P. aeruginosa, Bacillus, and Salmonella species from pharmaceutical preparations, raw materials, cosmetic samples, etc. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Pseudomonas aeruginosa - Colorless Escherichia coli - Pink-purple Staphylococcus aureus - Green to bluish green Salmonella Typhimurium - Colorless Salmonella Abony - Colorless

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Zhao et al.
Chemosphere, 263, 128344-128344 (2020-12-11)
The strategical integration of membrane water filtration with semiconductor photocatalysis presents a frontier in deep purification with a self-cleaning capability. However, the membrane fouling caused by the cake layer of the reclaimed TiO2 nanoparticles is a key obstacle. Herein, mesoporous
Kai-Wei Lin et al.
Bioorganic chemistry, 107, 104395-104395 (2021-01-02)
We report the design, synthesis, and biological evaluation of 13 new and 1 known anthraquinone derivatives which exerted cytotoxicity against PC3, A549 and NTUB1 cell lines. The results indicate that, among these 14, compounds-1 and 14 showed the highest growth
Yasamin Soleimanian et al.
Pharmaceutics, 12(4) (2020-04-26)
The objective of the present study was to explore the potential of nanostructured lipid carriers (NLCs) for improving the oral delivery of β-sitosterol, a poorly water-soluble bioactive component with hypocholesterolemic activity. Two β-sitosterol formulations with different solid lipid compositions were
Xuewu Zhu et al.
Environmental science & technology, 54(10), 6365-6374 (2020-04-24)
Traditional polyamide-based interfacial polymerized nanofiltration (NF) membranes exhibit upper bound features between water permeance and salt selectivity. Breaking the limits of the permeability and rejections of these composite NF membranes are highly desirable for water desalination. Herein, a high-performance NF
Corinna Braun et al.
Frontiers in cellular and infection microbiology, 10, 565808-565808 (2020-11-17)
Chlamydia pneumoniae is an obligate intracellular pathogen that causes diseases of the upper and lower respiratory tract and is linked to a number of severe and chronic conditions. Here, we describe a large, C. pneumoniae-specific cluster of 13 genes (termed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service