Skip to Content
Merck
All Photos(1)

Key Documents

70439

Millipore

Oxidase Test disks

suitable for agriculture, clinical testing, environmental, food and beverages pharmaceutical, microbiology and specific enzyme detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
eCl@ss:
32161000
NACRES:
NA.85

product name

Oxidase Test, suitable for microbiology

Agency

according to ISO 9308-1:2014

Quality Level

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Plesiomonas spp.
Aeromonas spp.
Neisseria spp.
Pseudomonas spp.
bacteria

Application

Oxidase Test is an important differential procedure which should be performed on all gram-negative bacteria that are to be identified.

Packaging

Discs contain N,N-dimethyl-p-phenylenediamine oxalate and α-naphthol.

Principle

Oxidase Test is based on the principle that certain bacteria produce indophenol blue from the oxidation of dimethyl-p-phenylenediamine and α-naphthol. In presence of the enzyme cytochrome oxidase (gram-negative bacteria), the N,N-dimethyl-p-phenylenediamine oxalate and α-naphthol react to indophenol blue.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baron S
Medical Microbiology null
Robin B Guevarra et al.
PloS one, 16(1), e0239792-e0239792 (2021-01-07)
Pantoea agglomerans is a Gram-negative facultative anaerobic bacillus causing a wide range of opportunistic infections in humans including septicemia, pneumonia, septic arthritis, wound infections and meningitis. To date, the determinants of virulence, antibiotic resistance, metabolic features conferring survival and host-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service