Skip to Content
Merck
All Photos(1)

Key Documents

00370580

Ginsenoside Rg1

primary reference standard

Synonym(s):

(3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-D-glucopyranoside, Ginsenoside A2, Ginsenoside g1, Panaxoside A, Panaxoside Rg1, Sanchinoside C1, Sanchinoside Rg1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12[C@@](C[C@H]([C@@]3([H])[C@]2(CC[C@]3([H])[C@@](CCC=C(C)C)(C)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO)C)O)([H])[C@@]5([C@@](C(C)([C@H](CC5)O)C)([H])[C@H](C1)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6O)O)O)CO)C

InChI

1S/C42H72O14/c1-20(2)10-9-13-42(8,56-37-34(52)32(50)30(48)25(19-44)55-37)21-11-15-40(6)28(21)22(45)16-26-39(5)14-12-27(46)38(3,4)35(39)23(17-41(26,40)7)53-36-33(51)31(49)29(47)24(18-43)54-36/h10,21-37,43-52H,9,11-19H2,1-8H3/t21-,22+,23-,24+,25+,26+,27-,28-,29+,30+,31-,32-,33+,34+,35-,36+,37-,39+,40+,41+,42-/m0/s1

InChI key

YURJSTAIMNSZAE-HHNZYBFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Takino
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 114(8), 550-564 (1994-08-01)
The pharmacodynamics of ginsenoside-Rg1 (Rg1), -Rb1 (Rb1) and -Rb2 (Rb2) in rats were studied. In these studies, obvious differences were found in their pharmacodynamics. That is, Rg1 was easily hydrated to the same prospogenins in both rat stomach and 0.1
Jiaying Wu et al.
Neurochemistry international, 62(1), 92-102 (2012-10-16)
Ginsenoside Rg1, a steroidal saponin of high abundance in ginseng, possesses the neuroprotective effects. In this study, we tried to explore the effect of Rg1 on promoting differentiation of mouse embryonic stem (ES) cells towards the neuronal lineage and its
Jung-Sun Cho et al.
Experimental biology and medicine (Maywood, N.J.), 237(6), 663-669 (2012-06-26)
Nasal polyps are associated with chronic inflammation of the sinonasal mucosa and are involved in myofibroblast differentiation and extracellular matrix (ECM) accumulation. Ginsenoside Rg1, a compound derived from Panax ginseng, shows antifibrotic and anticancer effects. However, the molecular effects of
Jin-Kwang Kim et al.
Journal of biotechnology, 161(3), 294-301 (2012-07-07)
This study focuses on the cloning, expression, and characterization of recombinant ginsenoside hydrolyzing glycosidase from Sanguibacter keddieii in order to biotransform ginsenosides efficiently. The gene, termed bglSk, consists of 1857 bp and revealed significant homology to that of glycoside hydrolase
Xiaoduo Guan et al.
Journal of pharmaceutical and biomedical analysis, 72, 261-266 (2012-09-27)
A simple and rapid high performance liquid chromatography electrospray ionization ion-trap tandem mass spectrometry (LC-MS/MS) method has been developed and validated for the quantitative determination of esculentoside A (EsA) in dog plasma using ginsenoside Rg1 as the internal standard (IS).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service