Skip to Content
Merck
All Photos(1)

Key Documents

00170580

Ginsenoside Rb1

primary reference standard

Synonym(s):

2-O-β-Glucopyranosyl-(3β,12β)-20-[(6-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl β-D-glucopyranoside, Arasaponin E1, Gynosaponin C, Gypenoside III, Notoginsenoside Rb1, Panaxoside Rb1, Sanchinoside E1, Sanchinoside Rb1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H92O23
CAS Number:
Molecular Weight:
1109.29
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

C\C(C)=C\CC[C@](C)(O[C@@H]1O[C@H](CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)[C@H]3CC[C@]4(C)[C@@H]3[C@H](O)C[C@@H]5[C@@]6(C)CC[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)C(C)(C)[C@@H]6CC[C@@]45C

InChI

1S/C54H92O23/c1-23(2)10-9-14-54(8,77-48-44(69)40(65)37(62)29(74-48)22-70-46-42(67)38(63)34(59)26(19-55)71-46)24-11-16-53(7)33(24)25(58)18-31-51(5)15-13-32(50(3,4)30(51)12-17-52(31,53)6)75-49-45(41(66)36(61)28(21-57)73-49)76-47-43(68)39(64)35(60)27(20-56)72-47/h10,24-49,55-69H,9,11-22H2,1-8H3/t24-,25+,26+,27+,28+,29+,30-,31+,32-,33-,34+,35+,36+,37+,38-,39-,40-,41-,42+,43+,44+,45+,46+,47-,48-,49-,51-,52+,53+,54-/m0/s1

InChI key

GZYPWOGIYAIIPV-JBDTYSNRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content measured by quantitative NMR can be found on the certificate.

Ginsenoside Rb1 is one of the primary constituents of traditional Chinese medicine, Ginseng. It belongs to the group of triterpene saponins called ginsenosides and pharmacologically, it is one of the most active dammarane-type ginsenosides. It is known to have anti-tumor, anti-oxidant, anti-inflammatory, and cytoprotective properties.

Application

The primary reference standard can be used for identification, assay, and purity testing, method development, and validation.It can also be used as follows:
  • Development and validation of a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the determination of ginsenoside Rb1, naringin, ginsenoside Rb2, and oridonin in rat plasma samples after administrating them weifuchun tablets
  • Multi-residue analysis of baicalin, baicalein, wogonoside, wogonin, scutellarin, berberine, coptisine, ginsenoside Rb1, and ginsenoside Re in biological samples of rat plasma by liquid chromatographic-tandem mass spectrometric (LC/MS/MS) method
  • Analysis of traditional Chinese herbal decoctions of Dushen Tang and Shenfu Tang for the quantification of 12 ginsenosides by UPLC-MS/MS technique
  • Analysis of rhizomes and roots of ginseng for the detection of 131 ginsenosides by UHPLC combined with a diode-array detector and quadrupole/time of flight tandem mass spectrometry (DAD-QTOF-MS/MS), along with the further determination of 19 ginsenosides by HPLC coupled with electrospray ionization-mass spectrometric detection

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiung-Hung Chang et al.
PloS one, 9(8), e105362-e105362 (2014-08-19)
Salmonella, a common zoonotic pathogen, causes gastroenteritis in both humans and animals. Traditional Chinese Medicine (TCM) has been used to improve gastrointestinal dysfunction and to modify the immune response to inflammation for centuries. This study used six herbal plants and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service