Skip to Content
Merck
All Photos(1)

Documents

37-0100

Sigma-Aldrich

Zinc

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 487 °C)

form

solid

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Pyr. Sol. 1 - Self-heat. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
I A Popov et al.
Molekuliarnaia biologiia, 47(3), 498-504 (2013-07-31)
Alzheimer's disease, is a lethal neurodegenerative pathology, characterized by the formation of soluble neurotoxic oligomers of the human amyloid-beta peptide Abeta which get accumulated forming polymeric extracellular aggregates (so-called amyloid plaques). The isomerized at aspartate 7 isoform of the human
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc
Marco Ranocchiari et al.
Chimia, 67(6), 397-402 (2013-08-16)
Structural features, synthesis, and reactivity of Zn-biphenyl metal-organic frameworks with MOF-5 topology are presented to show the chemical flexibility of such materials and to demonstrate the challenges that can be encountered and solved to avoid interpenetration. We introduce the synthesis
Cigdem Karakukcu et al.
Clinical laboratory, 59(5-6), 557-562 (2013-07-20)
The aim of this study was to assess the acute and chronic effects of vigorous physical activity, with and without zinc supplementation, on distribution of elements in young amateur boxers. The 8 week intervention trial experiment was designed with 32

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service