Skip to Content
Merck
All Photos(1)

Key Documents

33-0390

Sigma-Aldrich

Vinyl acetate

CP

Synonym(s):

Acetoxyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH=CH2
CAS Number:
Molecular Weight:
86.09
Beilstein:
1209327
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
CP
bp:
72-73 °C (lit.)
vapor pressure:
88 mmHg ( 20 °C)

grade

CP

vapor density

3 (vs air)

vapor pressure

88 mmHg ( 20 °C)

form

liquid

autoignition temp.

801 °F

expl. lim.

13.4 %

availability

available only in Japan

refractive index

n20/D 1.395 (lit.)

bp

72-73 °C (lit.)

mp

−93 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OC=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

17.6 °F

Flash Point(C)

-8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wlodarski et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 94, 106-115 (2015-05-23)
To improve solubility of tadalafil (Td), a poorly soluble drug substance (3μg/ml) belonging to the II class of the Biopharmaceutical Classification System, its six different solid dispersions (1:1, w/w) in the following polymers: HPMC, MC, PVP, PVP-VA, Kollicoat IR and
Anna Piperno et al.
The Journal of organic chemistry, 75(9), 2798-2805 (2010-04-17)
A new template of C-4'-truncated phosphonated nucleosides (TPCOANs) has been obtained in good yields according to two different routes which exploit the reactivity of a phosphonated nitrone. The one-step procedure based on the 1,3-dipolar cycloaddition of a phosphonated nitrone with
María B Blanco et al.
Environmental science & technology, 46(16), 8817-8825 (2012-07-18)
The products formed from the reactions of OH radicals with vinyl acetate and allyl acetate have been studied in a 1080 L quartz-glass chamber in the presence and absence of NO(x) using in situ FTIR spectroscopy to monitor the reactant
B Picquet-Varrault et al.
Environmental science & technology, 44(12), 4615-4621 (2010-05-25)
Vinyl acetate is widely used in industry. It has been classified as a high-production volume (HPV) chemical in the United States. To evaluate its impact on the environment and air quality, its atmospheric reactivity toward the three main tropospheric oxidants
Roberto M Risi et al.
Organic letters, 14(4), 1180-1182 (2012-02-07)
A highly atom-economical total synthesis of (+)-patulolide C has been accomplished in three steps from the known (2R)-8-nonyn-2-ol in 49% overall yield and 93% de. A Rh(I)-catalyzed asymmetric hydroformylation (AHF)/ intramolecular Wittig olefination cascade was utilized to set the C4-hydroxyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service