
Vinyl acetate

Synonym(s):
Acetoxyethylene, Vinyl acetate
Linear Formula:
CH3CO2CH=CH2
CAS Number:
Molecular Weight:
86.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing