Skip to Content
Merck
All Photos(1)

Key Documents

28-2950

Sigma-Aldrich

Sodium hydroxide

SAJ first grade, ≥95.0%

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥95.0%

form

solid

availability

available only in Japan

mp

318 °C (lit.)

solubility

water: ~1260 g/L at 20 °C

density

2.13 g/cm3

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • MOF-Enhanced Aluminosilicate Ceramic Membranes Using Non-Firing Processes for Pesticide Filtration and Phytochrome Removal.: This research utilizes sodium hydroxide in the development of MOF-enhanced aluminosilicate ceramic membranes. These membranes are crafted through non-firing processes, highlighting their utility in filtering pesticides and removing phytochromes, offering substantial benefits for environmental management and agricultural applications (Zhao L et al., 2024).

  • Assessing the Acidic and Alkaline Recalcitrance of Covalently Modified Surface Amines on Ordered Mesoporous Carbon.: The study examines the resistance of covalently modified surface amines on mesoporous carbon to acidic and alkaline environments, where sodium hydroxide is used to test alkaline recalcitrance. This work is pivotal for applications in catalysis and material science (Kovach NC et al., 2024).

  • Formation of plasmonic core/shell nanorods through ammonia-mediated dissolution of silver(i)oxide for ammonia monitoring.: This article discusses the role of sodium hydroxide in synthesizing plasmonic core/shell nanorods. The process involves ammonia-mediated dissolution, which is critical for developing sensors for ammonia detection in environmental monitoring (Ghorbanian E et al., 2024).

  • High temperature corrosion and oxide scale formation of nickel in molten NaOH at various basicity levels.: Investigates the corrosion behavior and oxide scale formation on nickel in molten sodium hydroxide. This study is crucial for understanding material degradation in high-basicity environments, which has implications for industrial processes involving caustic substances (Stoffersen B et al., 2024).

  • Long-term durability of discarded cork-based composites obtained by geopolymerization.: Focuses on the durability of cork-based composites treated with sodium hydroxide and geopolymerized. This research is significant for waste management and recycling processes, providing insights into sustainable material usage (Poggetto GD et al., 2024).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olof Idevall-Hagren et al.
The EMBO journal, 34(17), 2291-2305 (2015-07-24)
The extended synaptotagmins (E-Syts) are ER proteins that act as Ca(2+)-regulated tethers between the ER and the plasma membrane (PM) and have a putative role in lipid transport between the two membranes. Ca(2+) regulation of their tethering function, as well
Horacio Cardenas et al.
Epigenetics, 9(11), 1461-1472 (2014-12-04)
A key step in the process of metastasis is the epithelial-to-mesenchymal transition (EMT). We hypothesized that epigenetic mechanisms play a key role in EMT and to test this hypothesis we analyzed global and gene-specific changes in DNA methylation during TGF-β-induced
Aparajita H Chourasia et al.
EMBO reports, 16(9), 1145-1163 (2015-08-02)
BNip3 is a hypoxia-inducible protein that targets mitochondria for autophagosomal degradation. We report a novel tumor suppressor role for BNip3 in a clinically relevant mouse model of mammary tumorigenesis. BNip3 delays primary mammary tumor growth and progression by preventing the
Hélène O B Gautier et al.
Nature communications, 6, 8518-8518 (2015-10-07)
Myelin regeneration can occur spontaneously in demyelinating diseases such as multiple sclerosis (MS). However, the underlying mechanisms and causes of its frequent failure remain incompletely understood. Here we show, using an in-vivo remyelination model, that demyelinated axons are electrically active
Joshua S McLane et al.
Biophysical journal, 109(2), 249-264 (2015-07-23)
Mechanical properties of the tumor microenvironment have emerged as key factors in tumor progression. It has been proposed that increased tissue stiffness can transform stromal fibroblasts into carcinoma-associated fibroblasts. However, it is unclear whether the three to five times increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service