Skip to Content
Merck
All Photos(1)

Key Documents

24-2695

Sigma-Aldrich

Phosphorus standard solution

1000 ppm P

Synonym(s):

Potassium Phosphate Monobasic solution, Monopotassium phosphate, Potassium dihydrogen phosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid

grade

SAJ special grade

form

liquid

availability

available only in Japan

concentration

1000 ppm P

storage temp.

15-25°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ou Zhimin et al.
Applied biochemistry and biotechnology, 168(8), 2297-2308 (2012-10-17)
(S) -3-Chloro-1-(2-thienyl)-1-propanol was synthesized by the asymmetric reduction of 3-chloro-1-(2-thienyl)propanone with liquid-core immobilized Candida pseudotropicalis 104. The optimum time was 28 h for the re-cultivation of immobilized cells. The optimum film solvent for the liquid-core capsule was 0.3 % chitosan (M (w)
Mohd Shkir et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 103, 199-204 (2012-12-25)
Good quality single crystals of potassium dihydrogen orthophosphate (KDP) were grown with different doping concentration of Glycine by conventional solution technique in aqueous solution. X-ray diffraction study has been carried out in order to see the effect of dopant on
Jayasankar V Valiyaparambil et al.
The International journal of oral & maxillofacial implants, 27(5), 1271-1277 (2012-10-12)
To examine the relationship between dental cone beam computed tomography (CBCT) gray scale values and Hounsfield units (HU), and whether the gray values of edentulous sites correlate with the subjective clinical bone quality assessed at surgery. Two radiographic phantoms containing
Kamaleshwar P Singh et al.
Journal of industrial microbiology & biotechnology, 40(2), 235-244 (2012-11-28)
Secondary metabolites such as antibiotics are typically produced by actinomycetes as a response to growth limiting stress conditions. Several studies have shown that secondary metabolite production is correlated with changes observed in actinomycete pellet morphology. Therefore, we investigated the correlation
Kamran Ajmal Syed et al.
The Journal of chemical physics, 138(2), 024901-024901 (2013-01-17)
The efflorescence of an individual KH(2)PO(4) droplet on Teflon substrate was investigated by micro-Raman spectroscopy. With the decrease of relative humidity (RH) from 98.0% to 73.0%, the KH(2)PO(4) droplet lost water gradually and entered into supersaturated state, which was reflected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service