Skip to Content
Merck
All Photos(2)

Key Documents

531367

Sigma-Aldrich

Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)]

average Mw ~60,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H2O3 · C4H8)m · xH3N
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mw ~60,000

pH

7.0 (5% in H2O)

viscosity

10.3 cP, 0.5 % in H2O(25 °C)(lit.)

solubility

H2O: soluble

density

1.3 g/mL at 25 °C (lit.)

SMILES string

[N+H4].[N+H4].[O-]C(=O)\C=C\C(=O)[O-].O1C(=O)C=CC1=O.CC(=C)C

InChI

1S/C4H4O4.C4H2O3.C4H8.2H3N/c5-3(6)1-2-4(7)8;5-3-1-2-4(6)7-3;1-4(2)3;;/h1-2H,(H,5,6)(H,7,8);1-2H;1H2,2-3H3;2*1H3/b2-1+;;;;

InChI key

HJBFEUKGLFRPRQ-MBCFVHIPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)] (PIM) is a water soluble polymer.

Application

Binder for ceramic powders and dispersing agent for water soluble paints.
PIM may be used to prepare montmorillonite (MMT) clay and amylopectin-based nanocomposites. PIM may be used as a dispersant for the preparation of NiO/Y3O3-ZrO2 for solid oxide fuel cells (SOFCs). PIM may also find use as a coupling agent.Binder for ceramic powders and dispersing agent for water soluble paints.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Vasile & Gennadii Efremovich Zaikov
Environmentally Degradable Materials Based on Multicomponent Polymeric Systems null
Starch Nanocomposites with Platelet and Fibrillar Fillers.
Nordqvist D, et al.
Journal of Biobased Materials and Bioenergy, 3(2), 139-146 (2009)
Fabrication and Characterization of Anode-Supported Tubular Solid-Oxide Fuel Cells by Slip Casting and Dip Coating Techniques.
Zhang L, et al.
Journal of the American Ceramic Society. American Ceramic Society, 92(2), 302-310 (2009)
P B Kernoff et al.
Blood, 63(1), 31-41 (1984-01-01)
Circulating antibodies to factor VIII (anti-VIII, "inhibitors") occurring in patients with hemophilia neutralize porcine factor VIII less readily than human factor VIII in vitro. Over an 18-mo period, 8 patients with anti-VIII were treated with 45 courses (297 infusions) of
Abhimanyu Paraskar et al.
Nanotechnology, 22(26), 265101-265101 (2011-05-18)
The use of cisplatin, a first line chemotherapy for most cancers, is dose-limited due to nephrotoxicity. While this toxicity can be addressed through nanotechnology, previous attempts at engineering cisplatin nanoparticles have been limited by the impact on the potency of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service