Skip to Content
Merck
All Photos(1)

Documents

24-0200

Sigma-Aldrich

Pandy′s reagent

Synonym(s):

Phenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

in H2O

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Sanina et al.
FEBS letters, 587(14), 2260-2265 (2013-06-08)
The present work aimed to compare the effects of different lysophosphatidylethanolamine (LPE) content in lipids derived from Yersinia pseudotuberculosis cells exposed and not exposed to phenol on the conformation of OmpF-like porin of these bacteria. Differential scanning calorimetry and intrinsic
Kolishka V Tsekova et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 68(1-2), 53-59 (2013-05-11)
The capability of cells of the fungus Aspergillus awamori, either free or immobilized in hybrid sol-gel material cells, for phenol biodegradation was demonstrated. Phenol was present in the reaction mixture as the sole carbon and energy source, and its decomposition
Yanxue Han et al.
Journal of environmental management, 118, 196-204 (2013-03-05)
In this work, the surface structures of biochars, derived from three types of biomass, switchgrass (SG), hardwood (HW) and softwood (SW) through either fast pyrolysis (FP) in a fluidized-bed reactor (at 500 °C) or slow pyrolysis (at 500° and 700 °C), were
Amani M D Elahwany et al.
Acta biologica Hungarica, 64(1), 106-117 (2013-04-10)
Six isolates with phenol degrading ability were obtained from marine sediments by enrichment procedures and an isolate, AM4, was identified as Alcaligenes sp. by 16S rDNA sequencing. The Plackett-Burman design was applied to estimate the significance of culture medium components
B K Shormanov et al.
Sudebno-meditsinskaia ekspertiza, 56(2), 33-37 (2013-06-27)
The present work was designed to study the specific features of 2-methyl hydroxybezene and 3-methyl hydroxybenzene distribution after intragastric administration of these toxicants to warm-blooded animals (rats). They were detected in the unmetabolized form in the internal organs and blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service