Merck
Phenol biodegradation by fungal cells immobilized in sol-gel hybrids.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2013-05-11)
Kolishka V Tsekova, Georgi E Chernev, Alexander E Hristov, Lyudmila V Kabaivanova
ABSTRACT
The capability of cells of the fungus Aspergillus awamori, either free or immobilized in hybrid sol-gel material cells, for phenol biodegradation was demonstrated. Phenol was present in the reaction mixture as the sole carbon and energy source, and its decomposition was followed in repeated batch degradation experiments. Atomic force microscopy provided information on the development of self-organizing structures in the materials synthesized by the sol-gel method. Phenol biodegradation was mediated only by the fungal cells, and no absorption by the hybrid matrix was observed. Ten cycles of phenol biodegradation using the immobilized cells system were conducted during which up to 2000 mg l(-1) phenol was completely decomposed. Immobilized cells degraded phenol at 8.33 mg h(-1), twice as fast as free cells. The good performance of the immobilized fungal cell system is promising for the development of an efficient technology for treating phenol-containing waste waters.
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, ≥99.0%
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol stock solution, 100 mg/dL, standard
Sigma-Aldrich
Pandy′s reagent