Skip to Content
Merck
All Photos(1)

Documents

19-4060

Sigma-Aldrich

NMP

SAJ first grade, ≥98.0%

Synonym(s):

1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106420
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

product name

1-Methyl-2-pyrrolidinone, SAJ first grade, ≥98.0%

grade

SAJ first grade

vapor density

3.4 (vs air)

vapor pressure

0.29 mmHg ( 20 °C)
0.99 mmHg ( 40 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

518 °F

expl. lim.

9.5 %

availability

available only in Japan

refractive index

n20/D 1.47 (lit.)

pH

7.7-8

bp

202 °C (lit.)
81-82 °C/10 mmHg (lit.)

mp

−24 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1=O

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3

InChI key

SECXISVLQFMRJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Vogelaar et al.
Small (Weinheim an der Bergstrasse, Germany), 1(6), 645-655 (2006-12-29)
Phase separation micromolding (PSmicroM) is a versatile microfabrication technique that can be used to structure a very broad range of polymers, including block copolymers and biodegradable and conductive polymers without the need for clean-room facilities. By incorporating a subsequent process
Hsiao-Chun Wang et al.
European journal of medicinal chemistry, 84, 312-334 (2014-07-19)
Bioisosteric replacement of acylureido moiety in 6-acylureido-3-pyrrolylmethylidene-2-oxoindoline derivatives resulted in a series of malonamido derivatives with indolin-2-one scaffold (11-14). Further conformational restrictions of the malonamido moiety led to 2-oxo-1,2-dihydropyridine (21-25) or a 4-oxo-1,4-dihydropyridine derivatives (31-36). 4-Oxo-1,4-dihydropyridine derivatives were more potent
Ajit Dhananjay Jagtap et al.
European journal of medicinal chemistry, 85, 268-288 (2014-08-05)
A series of 6-acylureido derivatives containing a 3-(pyrrol-2-ylmethylidene)indolin-2-one scaffold were synthesized as potential dual Aurora B/FLT3 inhibitors by replacing the 6-arylureido moiety in 6-arylureidoindolin-2-one-based multi-kinase inhibitors. (Z)-N-(2-(pyrrolidin-1-yl)ethyl)-5-((6-(3-(2-fluoro-4-methoxybenzoyl)ureido)-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide (54) was identified as a dual Aurora B/FLT3 inhibitor (IC50 = 0.4 nM and 0.5 nM, respectively).
Nurit Atar et al.
ACS applied materials & interfaces, 7(22), 12047-12056 (2015-05-07)
In low Earth orbit (LEO), hazards such as atomic oxygen (AO) or electrostatic discharge (ESD) degrade polymeric materials, specifically, the extensively used polyimide (PI) Kapton. We prepared PI-based nanocomposite films that show both AO durability and ESD protection by incorporating
Yichao Wang et al.
Nano letters, 15(2), 883-890 (2015-01-07)
The exhibition of plasmon resonances in two-dimensional (2D) semiconductor compounds is desirable for many applications. Here, by electrochemically intercalating lithium into 2D molybdenum disulfide (MoS2) nanoflakes, plasmon resonances in the visible and near UV wavelength ranges are achieved. These plasmon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service