Skip to Content
Merck
All Photos(1)

Key Documents

13-1520

Sigma-Aldrich

Hydrazine monohydrate

SAJ first grade, ≥98.0%

Synonym(s):

Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2O
CAS Number:
Molecular Weight:
50.06
Beilstein:
878137
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 25 °C)

Assay

≥98.0%

form

liquid

expl. lim.

99.99 %

availability

available only in Japan

refractive index

n20/D 1.428 (lit.)

pH

10.6-10.7 (10 g/L)

bp

120.1 °C (lit.)

mp

−51.7 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

O.NN

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boran Kartal et al.
FEMS microbiology reviews, 37(3), 428-461 (2012-12-06)
Anaerobic ammonium-oxidizing (anammox) bacteria primarily grow by the oxidation of ammonium coupled to nitrite reduction, using CO2 as the sole carbon source. Although they were neglected for a long time, anammox bacteria are encountered in an enormous species (micro)diversity in
Weidong Qu et al.
Biosensors & bioelectronics, 42, 430-433 (2012-12-19)
Graphene-nickel nanoparticle hybrid was prepared by the one-step far infrared-assisted reduction of graphene oxide and nickel (II) ions using hydrazine. It was loaded on the surface of a magnetic electrode for electrochemical sensing. The feasibility and performance of the novel
Timothy M Alligrant et al.
Lab on a chip, 13(3), 349-354 (2012-12-06)
We report on real-time electrochemical detection of individual DNA hybridization events at an electrode surface. The experiment is carried out in a microelectrochemical device configured with a working electrode modified with single-stranded DNA probe molecules. When a complementary DNA strand
Mild and rapid Pd-catalyzed cross-coupling with hydrazine in continuous flow: application to the synthesis of functionalized heterocycles.
Andrew DeAngelis et al.
Angewandte Chemie (International ed. in English), 52(12), 3434-3437 (2013-02-14)
Erika Lattová et al.
Mass spectrometry reviews, 32(5), 366-385 (2013-01-25)
Over the last years, extensive studies have evaluated glycans from different biological samples and validated the importance of glycosylation as one of the most important post-translational modifications of proteins. Although a number of new methods for carbohydrate analysis have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service