Merck
Electrochemical detection of individual DNA hybridization events.
Lab on a chip (2012-12-06)
Timothy M Alligrant, Elizabeth G Nettleton, Richard M Crooks
ABSTRACT
We report on real-time electrochemical detection of individual DNA hybridization events at an electrode surface. The experiment is carried out in a microelectrochemical device configured with a working electrode modified with single-stranded DNA probe molecules. When a complementary DNA strand labelled with a catalyst hybridizes to the probe, an easily detectable electrocatalytic current is observed. In the experiments reported here, the catalyst is a platinum nanoparticle and the current arises from electrocatalytic oxidation of hydrazine. Two types of current transients are observed: short bursts and longer-lived steps. At low concentrations of hydrazine, the average size of the current transients is proportional to the amount of hydrazine present, but at higher concentrations the hydrazine oxidation reaction interferes with hybridization.
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine, anhydrous, 98%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazinium dichloride, SAJ special grade, ≥98.0%
Sigma-Aldrich
Hydrazine monohydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol