Skip to Content
Merck
All Photos(1)

Key Documents

09-2080

Sigma-Aldrich

Ethyl lactate

SAJ first grade, ≥97.5%

Synonym(s):

Ethyl 2-hydroxypropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOCH2CH3
CAS Number:
Molecular Weight:
118.13
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

Assay

≥97.5%

availability

available only in Japan

refractive index

n20/D 1.413 (lit.)

bp

154 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)O

InChI

1S/C5H10O3/c1-3-8-5(7)4(2)6/h4,6H,3H2,1-2H3

InChI key

LZCLXQDLBQLTDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabetta Ranucci et al.
Macromolecular bioscience, 4(8), 706-713 (2004-10-07)
1-Vinyl-2-pyrrolidinone (VP) oligomers bearing a lactate group at one end (PVP-L) were obtained by chain-transfer controlled radical polymerisation carried out in the presence of ethyl L-lactate as chain-transfer agent (CTA). Their number-average molecular weights were in the range 1500-4000 with
Jun Liu et al.
The Journal of organic chemistry, 77(21), 9718-9723 (2012-10-12)
The efficient and concise synthesis of (-)-orthodiffenes A and C has been accomplished for the first time in eight steps from readily available chiral synthons, D-mannose and D-ethyl lactate. Our work confirmed the complete structure of orthodiffenes A and C
Chiaki Inaba et al.
Applied microbiology and biotechnology, 83(5), 859-864 (2009-03-17)
The whole-cell biocatalyst displaying Candida antarctica lipase B (CALB) on the yeast cell surface with alpha-agglutinin as the anchor protein was easy to handle and possessed high stability. The lyophilized CALB-displaying yeasts showed their original hydrolytic activity and were applied
O Dudeck et al.
AJNR. American journal of neuroradiology, 27(9), 1900-1906 (2006-10-13)
The organic solvent dimethyl-sulfoxide (DMSO), as a commonly used vehicle for nonadhesive liquid embolics, is not devoid of local angiotoxic effects. We compared microvascular toxicities of superselective infusions of DMSO with potentially more compatible solvents in swine rete mirabile. Fourteen
Hongyan Guo et al.
Journal of hazardous materials, 174(1-3), 59-63 (2009-09-29)
A new approach using aqueous ethyl lactate-modified [S,S]-ethylenediaminedisuccinic acid (EDDS) washing solutions for the removal of Cu from field-contaminated soils was examined in the laboratory. At pH 7 the maximum extraction efficiencies of Cu were about 41% by EDDS solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service