Skip to Content
Merck
All Photos(1)

Key Documents

35-0100

Sigma-Aldrich

Xylenes

SAJ special grade, ≥80.0%

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ special grade

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥80.0%

form

liquid

expl. lim.

7 %

availability

available only in Japan

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Vapour-Liquid Equilibrium for Xylenes + Tetrapropylene Glycol Binary Systems: Experimental Data and Regression.: This study presents experimental data and regression analyses for vapour-liquid equilibrium in xylenes and tetrapropylene glycol binary systems, providing valuable insights for separation processes in chemical engineering (Fendu et al., 2022).



Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-France le Rolle et al.
Journal of translational medicine, 13, 199-199 (2015-06-25)
Clinical therapy for metastatic colorectal cancer (CRC) remains limited, especially when the tumor harbors a KRAS mutation. This study aimed to identify prognostic biomarkers in CRC that are accessible for therapeutic inhibition. Conditioned media from human CRC epithelial cells and
Ryan P Smith et al.
Human gene therapy, 26(6), 377-385 (2015-03-27)
Much research has gone into the development of hybrid gene delivery systems that combine the broad tropism and efficient transduction of adenoviral vectors with the ability to achieve stable expression of cargo genes. In addition to gene therapy applications, such
Osceola Whitney et al.
Developmental neurobiology, 75(7), 778-790 (2014-11-20)
The forkhead domain FOXP2 and FOXP1 transcription factors are implicated in several cognitive disorders with language deficits, notably autism, and thus play a central role in learned vocal motor behavior in humans. Although a similar role for FoxP2 and FoxP1
Chani M Becker et al.
Neuro-oncology, 17(9), 1210-1219 (2015-05-15)
Targeting drug delivery to invasive glioma cells is a particularly difficult challenge because these cells lie behind an intact blood-brain barrier (BBB) that can be observed using multimodality imaging. BBB-associated efflux transporters such as P-glycoprotein (P-gp) and breast cancer resistance
Lian He et al.
Journal of environmental sciences (China), 23(1), 160-165 (2011-04-12)
The activities of CeO2 nanocubes calcined at different temperatures were tested for catalytic oxidation of o-xylene. Using CeO2 nanocubes as catalysts, complete catalytic oxidation of o-xylene was achieved below 210 degrees C. The CeO2 nanomaterials were characterized by means of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service