11051458001
Roche
dCTP
=98%, solution, 100 mM, suitable for DNA sequencing, suitable for DNA amplification
Synonym(s):
dCTP, 2′-deoxy-cytidine-5′-triphosphate, deoxy-cytidine-5′-triphosphate, 2′-
About This Item
Recommended Products
Quality Level
description
Lithium salt
Assay
98%
form
solution
usage
sufficient for 6,250 standard reactions (PCR)
packaging
pkg of 250 μL (25 μmol)
manufacturer/tradename
Roche
concentration
100 mM
technique(s)
DNA amplification: suitable
DNA sequencing: suitable
storage temp.
−20°C
Related Categories
General description
dCTP solution in water (pH 7), 100 mM, lithium salt.
Application
Quality
- ≥98% dCTP (HPLC), ≤1.5% dCDP (HPLC), ≤0.5% dCMP (HPLC)
- Absorbance determination A250/A260, A280/A260, A290/A260
- DNase activity negative, according to the current Quality Control procedures
- Suitability for PCR
Unit Definition
Storage and Stability
Note: Avoid repeated freezing and thawing. Store in aliquots.
Other Notes
Our PCR Grade deoxynucleotides are even purer than the lithium salts, to ensure excellent performance in your PCRs. PCR Grade deoxynucleotides contain the sodium salt of the nucleotide in PCR Grade Water.
For your convenience, some of our deoxynucleotides are also available in:
- nucleotide sets (all 4 nucleotides in separate vials)
- ready-to-use nucleotide mixes (premixed solutions of all 4 nucleotides)
They are also vital components of:
- kits (dNTPs, buffer, and polymerase in separate vials)
- ready-to-use mixes [polymerase, nucleotides, and buffer (premixed 2x solutions)]
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service